Learn More
World Precision Instrument Microelectrode Holder (MEH3S)-MEH3S15
SDP
Supplier:  World Precision Instrument MEH3S15
Encompass
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. Please specify diameter of glass you are using when placing your order.
Catalog No. 50-252-1114