Learn More
Research Products International Corp MICROFLEX 92-134 Gloves, Extra Small, 100/bx
SDP
Supplier:  Research Products International Corp 036530XS
Encompass_Preferred
The MICROFLEX 92-134 disposable nitrile examination glove offers tactility and comfortable protection across numerous applications. The lightweight, soft and stretchy nitrile formulation offers a comfortable fit and feel with the tactile sensitivity needed for fine motor control. Designed with textured fingertips for secure handling of tools and materials.Material: Nitrile Not Made From Natural Rubber Latex: YesColor: Blue Cuff Length: Standard Powder Content: Powder-Free External Glove Surface: Textured Fingers Freedom from Holes (INSPECTION LEVEL I): 1.5 AQL Palm Thickness (MM/MIL): 0.07 / 2.8Finger Thickness (MM/MIL): 0.11 / 4.3Allergy Prevention: Latex (TYPE I)Available Sizes: XS (5.5 - 6), S (6.5 - 7), M (7.5 - 8), L (8.5 - 9), XL (9.5 - 10) Research or further manufacturing use only, not for food or drug use.
Catalog No. 50-225-3006
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.