Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ ApexPro™ XP100 APXP12 Nitrile Gloves

Catalog No. 19500942 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Medium
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-942 X-Large
19-500-938 X-Small
19-500-940 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-500-942 Supplier Ansell Supplier No. APXP124

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual layer, disposable nitrile exam glove with extended cuff provides protection against opioids and other substances. Black exterior and grey interior make hazardous materials more visible. Fully textured design makes this glove applicable in a wide variety of healthcare, laboratory and industrial industries.

  • Black exterior and gray interior to make potentially hazardous powders more visible
  • Tested against both fentanyl and gastric acid
  • Dual layer, dual color design
  • Advanced barrier protection (AQL 0.65)
  • pH balanced interior coating for improved hand health
  • Recommended for a variety of industries including:
    • Chemical
    • Correctional Facilities
    • Emergency Medical Services
    • Healthcare
    • Law Enforcement
    • Life Sciences
    • Machinery and Equipment
    • Maintenance
    • Recycling and Waste Disposal
    • Transportation
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319; FDA21 CFR 177-2600; ISO 13485; ISO 9001; NFPA 1999:2018
Chemotherapeutic Rated No
Applications Healthcare, Industrial, Laboratory
AQL 0.65
Color Black, Gray
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.21 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.9 mil
Palm Thickness (Metric) 0.15 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Thickness (English) 5.9 mil
Thickness (Metric) 0.15 mm
FDA 510 (k) No
Model APXP12-4
Quantity 1000/Cs.
Series MICROFLEX™ ApexPro™ XP100 APXP12
Tensile Strength 22 MPa
Length (English) 11.8 in.
Length (Metric) 29.9 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.