Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ Lifestar EC™ LSE-104 High Risk Nitrile Gloves
GREENER_CHOICE

Catalog No. 19010315 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-315 Small
19-010-316 Medium
19-010-317 Large
19-010-318 X-Large
19-010-319 2X-Large
19-010-320 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-315 Supplier Ansell Supplier No. LSE104S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disposable EMS gloves provide protection against bloodborne pathogens, chemical splashes and other hazardous substances. Dual layer, dual color, high risk exam gloves are specially designed for use while caring for patients, administering drugs, and performing other vital tasks.

  • Designed specifically for EMS and healthcare professionals
  • Durable for safe critical patient care
  • Dual layer, dual color design for two layers of protection
  • Bright blue interior makes breaches more detectable
  • White exterior makes fluid and blood contact more visible
  • Tested against both fentanyl and gastric acid to simulate hazardous, real world overdose situations
  • Advanced barrier protection (AQL 0.65)
  • Non-stick and non-foaming properties for reduced interference when working
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600, ISO 13485, ISO 9001, NFPA 1999:2013
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Chlorinated Yes
Accelerator Free Yes
Applications Healthcare
AQL 0.65
Color White, Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.21 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
FDA 510 (k) No
Glove Cuff Style Beaded
Model LSE-104-S
Quantity 1000/Cs.
Series MICROFLEX™ Lifestar EC™ LSE-104
Tensile Strength 25 MPa
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.