Learn More
Description
- Designed specifically for EMS and healthcare professionals
- Durable for safe critical patient care
- Dual layer, dual color design for two layers of protection
- Bright blue interior makes breaches more detectable
- White exterior makes fluid and blood contact more visible
- Tested against both fentanyl and gastric acid to simulate hazardous, real world overdose situations
- Advanced barrier protection (AQL 0.65)
- Non-stick and non-foaming properties for reduced interference when working
Specifications
Specifications
| Certifications/Compliance | ASTM D6319, FDA21 CFR 177-2600, ISO 13485, ISO 9001, NFPA 1999:2013 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Chemotherapeutic Rated | No |
| Applications | Healthcare |
| AQL | 0.65 |
| Color | White, Blue |
| Disposable | Single-use |
| Exterior Finish | Partially Textured |
| Finger Thickness (English) | 8.3 mil |
| Finger Thickness (Metric) | 0.21 mm |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.

