Learn More
Microflex™ Micro One™ Latex Exam Gloves
Catalog No. 19044470 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-044470 Medium
18-044-471 Large
19-044469 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-044470 Supplier Microflex™ Supplier No. MO150M
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Formulated for proper comfort, fit and feel
  • Lightly powdered examination gloves feature smooth, nontextured surface
  • Interior is coated with absorbable USP cornstarch
  • Test values measure below proposed Federal quality and performance guidelines for maximum residual powder and protein content
  • Thickness is 6.3mil. minimum at fingertip
  • Beaded cuffs
  • Nonsterile
  • Ambidextrous
  • Single-use
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D 3578
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powdered
Size Medium
Sterility Non-sterile
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
Length (English) 9.5 in.
Length (Metric) 24.1 cm