Learn More
International Enviroguard™ MicroGuard MP Microporous Frock with Mandarin Collar, No Pockets
Shop All International Enviroguard Systems ProductsDescription
- Fabric keeps dust and liquids out while allowing air and moisture to flow freely, so wearers stay cool throughout a hard day's work
- Ideal for non-hazardous work environments that require reliable liquid and particulate protection
- Frocks are produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
- Frock does not have pockets
- Secure, latex-free elastic wrists help keep sleeves in place to prevent skin exposure
- Front snap closures are easy to secure
- Low-linting fabric helps prevent cross-contamination
- Possible applications include food processing, industrial applications, pharmaceuticals, cleanrooms and lab applications, and more
Specifications
Specifications
| Size | 5X-Large |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snap (Front) |
| Cuff Style | Elastic |
| Collar Type | Mandarin |
| Latex Free | Yes |
| Sleeve Style | Long Sleeve |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.