Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard MP Microporous Frock with Mandarin Collar, No Pockets

Catalog No. 17184076 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-076 4X-Large
17-184-074 Large
17-184-073 Medium
17-184-072 X-Large
17-184-078 2X-Large
17-184-077 3X-Large
17-184-075 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-184-076 Supplier International Enviroguard™ Supplier No. 80484XL
Add to Cart
Add to Cart

Microporous Frock with elastic wrists and front snap closure offers the ideal balance of safety and comfort.

  • Fabric keeps dust and liquids out while allowing air and moisture to flow freely, so wearers stay cool throughout a hard day's work
  • Ideal for non-hazardous work environments that require reliable liquid and particulate protection
  • Frocks are produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
  • Frock does not have pockets
  • Secure, latex-free elastic wrists help keep sleeves in place to prevent skin exposure
  • Front snap closures are easy to secure
  • Low-linting fabric helps prevent cross-contamination
  • Possible applications include food processing, industrial applications, pharmaceuticals, cleanrooms and lab applications, and more

Specifications

Size 4X-Large
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snap (Front)
Cuff Style Elastic
Collar Type Mandarin
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable
Product Type Frock
Seam Type Serged
For Use With (Application) Keeping Out Dust and Liquids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.