Learn More
REPLIGEN CORPORATION Repligen MicroKros Hollow Fiber Module, 100 kD MWCO, Sterile Condition, 1 Unit
SDP
Supplier:  REPLIGEN CORPORATION C06E10005N
Encompass
The Repligen MicroKros is a versatile 65 cm hollow fiber filter module designed for efficient tangential flow filtration in biological applications. With its 100 kD molecular weight cut-off and mPES membrane, it excels in concentration, diafiltration, and purification processes. The compact design with a 0.50 mm fiber inner diameter promotes seamless integration into lab setups, ensuring high-quality results in sensitive research.
Catalog No. NC2160674
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.