Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ MicroLiter™ 11 mm Crimp Top Autosampler Vial Seals
SDP

Catalog No. 15111486 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Gold
Green
Red
Silver
Septum:
ETFE, Silicone
FEP, Butyl
FEP, Natural Rubber
PTFE, Silicone
PTFE, Silicone, PTFE
Quantity:
100 Caps/Pk.
2000 Caps/Pk.
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Quantity
15-111-486 Blue FEP, Natural Rubber 100 Caps/Pk.
15-111-518 Silver PTFE, Silicone 2000 Caps/Pk.
15-111-524 Silver PTFE, Silicone, PTFE 2000 Caps/Pk.
15-111-483 Silver FEP, Natural Rubber 100 Caps/Pk.
15-111-484 Silver FEP, Natural Rubber 2000 Caps/Pk.
15-111-517 Silver PTFE, Silicone 100 Caps/Pk.
15-111-523 Silver PTFE, Silicone, PTFE 100 Caps/Pk.
15-111-509 Blue ETFE, Silicone 100 Caps/Pk.
15-111-491 Red FEP, Natural Rubber 100 Caps/Pk.
15-111-513 Red ETFE, Silicone 100 Caps/Pk.
15-111-511 Green ETFE, Silicone 100 Caps/Pk.
01-549-776 Green PTFE, Silicone 100 Caps/Pk.
15-111-522 Gold PTFE, Silicone 100 Caps/Pk.
15-111-495 Silver FEP, Butyl 100 Caps/Pk.
15-111-506 Silver ETFE, Silicone 100 Caps/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 15-111-486 Supplier DWK Life Sciences™ Supplier No. 110020B
Add to Cart
Add to Cart

MicroLiter™'s 11 mm, crimp top autosampler vial seals are available in multiple colors and septa types.

  • Crimp top vials specific to the MicroLiter™ autosampler
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Aluminum
Pre-slit No
Septum FEP, Natural Rubber
Product Line MicroLiter
Quantity 100 Caps/Pk.
Closure Size 11 mm
For Use With (Equipment) Autosampler Vials
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.