Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111724 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Aluminum, Silicone
Butyl Stopper
Chemically Inert Butyl
Natural PTFE, Blue Silicone
PTFE, Butyl
PTFE, Molded Butyl
Tan PTFE, White Silicone
No. per Pack:
100
1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum No. per Pack
15-111-724 Tan PTFE, White Silicone 1000
15-111-658 Chemically Inert Butyl 100
15-111-673 Butyl Stopper 100
15-111-678 PTFE, Butyl 100
15-111-691 PTFE, Molded Butyl 100
15-111-703 Natural PTFE, Blue Silicone 100
15-111-723 Tan PTFE, White Silicone 100
15-111-740 Aluminum, Silicone 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15-111-724 Supplier DWK Life Sciences Supplier No. 200051ASB
Add to Cart
Add to Cart

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Material Aluminum
Pre-slit No
Septum Tan PTFE, White Silicone
No. per Pack 1000
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.