Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Bi-Metal Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111681 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Septum:
Aluminum, Silicone
Molded PTFE, Butyl
Natural PTFE, Blue Silicone
None
PTFE, Butyl
Tan PTFE, White Silicone
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15-111-681 Blue PTFE, Butyl
15-111-631 Green None
15-111-706 Blue Natural PTFE, Blue Silicone
15-111-707 Green Natural PTFE, Blue Silicone
15-111-708 Red Natural PTFE, Blue Silicone
15-111-727 Blue Tan PTFE, White Silicone
15-111-728 Green Tan PTFE, White Silicone
15-111-745 Red Aluminum, Silicone
15-111-630 Blue None
15-111-694 Blue Molded PTFE, Butyl
15-111-729 Red Tan PTFE, White Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 15-111-681 Supplier DWK Life Sciences Supplier No. 200030BMB
Add to Cart
Add to Cart

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Steel
Pre-slit No
Septum PTFE, Butyl
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.