Learn More
DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers
Shop All DWK Life Sciences Products

Description
Specifications
Specifications
| Color | Blue, Natural |
| Material | PTFE, Silicone |
| Pre-slit | No |
| No. per Pack | 100 |
| Closure Size | 20 mm |
| For Use With (Equipment) | 20 mm Headspace Seals - MicroLiter |
| Product Type | Septa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.