Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20mm Magnetic Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111730 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gold
Silver
Material (Liner):
Aluminum/Silicone
Chemically Inert Butyl
Natural PTFE/Blue Silicone
PTFE/Butyl
PTFE/Molded Butyl
Tan PTFE/White silicone
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material (Liner)
15-111-730 Silver Tan PTFE/White silicone
15-111-684 Silver PTFE/Butyl
15-111-697 Silver PTFE/Molded Butyl
15-111-711 Silver Natural PTFE/Blue Silicone
15-111-716 Gold Natural PTFE/Blue Silicone
15-111-735 Gold Tan PTFE/White silicone
15-111-667 Gold Chemically Inert Butyl
15-111-747 Gold Aluminum/Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15-111-730 Supplier DWK Life Sciences Supplier No. 200051M
Add to Cart
Add to Cart

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

Magnetic crimp seal
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Tan PTFE/White silicone
Color Silver
Material Steel
Pre-slit No
Septum PTFE/Silicone
Quantity 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.