Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with ETFE/Silicone Septa
SDP

Catalog No. 15111061 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
Product Type:
Screw Cap
Quantity:
100
2000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity
15-111-061 Red Screw Cap 2000
15-111-050 Blue Screw Cap 100
15-111-051 Blue Screw Cap 2000
15-111-053 Green Screw Cap 100
15-111-055 Black Screw Cap 100
15-111-057 Natural Screw Cap 100
15-111-060 Red Screw Cap 100
15-111-062 Yellow Screw Cap 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15-111-061 Supplier DWK Life Sciences Supplier No. 090022RB
Add to Cart
Add to Cart

Available in multiple colors and package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) ETFE/Burnt Red Silicone
Color Red
Material Polypropylene
Pre-slit No
Septum ETFE/Silicone
Quantity 2000
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.