Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial Caps with FEP/Natural Rubber Septa
SDP

Catalog No. 15111028 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
No. per Pack:
100
2000
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15-111-028 Red 100
15-111-016 Blue 100
15-111-017 Blue 2000
15-111-019 Green 100
15-111-020 Green 2000
15-111-023 Black 2000
15-111-026 Natural 2000
15-111-029 Red 2000
15-111-031 Yellow 100
15-111-032 Yellow 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 15-111-028 Supplier DWK Life Sciences Supplier No. 090020R
Add to Cart
Add to Cart

Available in multiple colors and package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polypropylene
Pre-slit No
Septum FEP, Natural Rubber, FEP
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.