Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Septa
SDP

Catalog No. 15111065 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Orange
Red
Red, White
White
White, Blue
Septum:
ETFE/Silicone
FEP/Natural Rubber
PTFE
PTFE/Silicone
PTFE/Silicone/PTFE
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15-111-065 Red, White PTFE/Silicone
15-111-049 Orange ETFE/Silicone
15-111-014 White PTFE
15-111-015 Orange FEP/Natural Rubber
15-111-085 White, Blue PTFE/Silicone
15-111-103 Red PTFE/Silicone/PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-111-065 Supplier DWK Life Sciences Supplier No. 090030
Add to Cart
Add to Cart

MicroLiter septa for 9mm Screwthread Autosampler Vials are constructed with a wide array of chemical compatibilities to suit a variety of chromatography applications.

TRUSTED_SUSTAINABILITY

Specifications

Color Red, White
Pre-slit No
Septum PTFE/Silicone
Quantity 100
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.