Learn More
DWK Life Sciences MicroLiter Glass Limited Volume Inserts for 11 mm Autosampler Vials, Untreated
Shop All DWK Life Sciences Products

Specifications
Specifications
| Color | Clear |
| Material | Borosilicate Glass (Type 1) |
| No. per Pack | 100 |
| Volume (Metric) Insert | 350 μL |
| Base Style | Flat Bottom |
| Product Line | MicroLiter |
| For Use With (Equipment) | 11 mm Autosampler Vials - MicroLiter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.