Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Catalog No. 2821 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
28-21 -
14-070-88 HM 550 and 560
52-449-0 HM 525
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-21 Supplier Epredia™ Supplier No. 2821
Add to Cart
Add to Cart
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Block surface planed parallel to knife edge to minimize trimming
  • Handheld to permit use in all makes of cryotomes and cryostats
  • Stainless-steel block is 1.2in. (3cm) diameter
  • Steel stem is 3in. (7.6cm) long and fitted with a plastic knob
Specifications
Product Type Heat Extractor