Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038528 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Gray
Green
Lilac
Orange
Peach
Pink
Tan
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-528 Aqua
22-038-519 Pink
22-038-520 Green
22-038-523 Peach
22-038-524 Tan
22-038-525 Gray
22-038-526 Lilac
22-038-527 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-038-528 Supplier Simport™ Scientific Supplier No. M50712T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks. No need for biopsy pads thanks to the 1676 square 0.36 mm openings of the mesh.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • 1676 square opening (0.36 mm)
  • No biopsy pads needed
  • 1 large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Aqua
Material Acetal