Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes
Catalog No. 23045374 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Peach
Tan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-374 Peach
23-045-375 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-374 Supplier Simport™ Scientific Supplier No. M5077
Add to Cart
Add to Cart
This item is not returnable. View return policy
Micromesh™ Biopsy Processing/Embedding Cassettes are molded from a special high density polymer and are designed to keep specimens safely submerged in liquid.
  • Totally resistant to the chemical action of histological solvents.
  • Lid can be opened and closed as often as necessary and always re-locks securely without danger of specimen loss.
  • Offers 1,676 square openings (0.38 mm) allowing for a greatly improved fluid exchange and drainage without having to use biopsy pads.
  • Large anterior slots in both cassette and cover ensure that the Micromesh™ biopsy cassette will sink rapidly.
  • Large square compartment measuring 27 mm is ideal even for needle biopsies.
  • Cover does not protrude above the cassette, a space saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
  • Secure locking device safely holds the specimen during processing.
  • 4.22 mm inside height.
Specifications
Quantity 250/Box, 4 Boxes/Cs.
Packaging Type Dispenser Boxes
Writing Face Angle 45°
Color Peach
Material Acetal
Product Type Cassette