Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes with 4 Compartments
Catalog No. 23045376 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Gray
Peach
Tan
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-376 Peach
23-045-377 Tan
23-045-378 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-045-376 Supplier Simport™ Scientific Supplier No. M5087
Add to Cart
Add to Cart
This item is not returnable. View return policy
Micromesh™ Biopsy Processing/Embedding Cassettes with 4 Compartments are designed so users do not need to use biopsy pads.
  • Offers 1,676 square openings (0.38 mm) allowing for a greatly improved fluid exchange and drainage without having to use biopsy pads.
  • Four square compartments, each measuring 13 mm.
  • Large anterior slots in both cassette and cover ensure that the Micromesh™ biopsy cassette will sink rapidly.
  • Cover does not protrude above the cassette, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
  • Secure locking device safely holds the specimen during processing.
Specifications
Quantity 250/Box, 4 Boxes/Cs.
Packaging Type Dispenser Boxes
Writing Face Angle 45°
Color Peach
Material Acetal
Product Type Cassette