Learn More
Ajax Scientific Micrometer Caliper
SCI_ED
Catalog No. S40505
Click to view available options
Measures in metric units; Screw pitch: 0.5mm; Least count: 0.01mm
Catalog No.
S40505 Measures in metric units; Screw pitch: 0.5mm; Least count: 0.01mm
1 options
This item is not returnable. View return policy
Catalog No. S40505 Supplier Ajax Scientific Supplier No. 12078
Add to Cart
Add to Cart
This item is not returnable. View return policy
Traditional English caliper with a 0-1 in. range
Calipers show decimal equivalents and have a satin finish with friction stop. Graduations on the English-Unit caliper are 0.001 in.. Metric Caliper has a 0-25mm range with 0.01mm graduations. Spindle locks for more precise comparison.
Specifications
Product Type Metric Unit Caliper
Includes Hardened Steel screw, zero adjustment and locking device