Learn More
Glass Expansion MicroMist DC Nebulizer 0.1mL/min, Includes NFT-025 UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (1 piece) and 70-803-0969 Gas Fitting with Ratchet Nebulizer connector
SDP
Supplier:  Glass Expansion A1107UM01
Encompass_Preferred
MicroMist DC Nebulizer 0.1 mL/min, includes UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (1 pc.) and DC Gas Fitting Type 11 with ratchet connector (1 pc.). Compatible with Vista/700-ES Series ICP-OES, 4100/4200 MP-AES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 0.1mL/min. Recommended not to exceed optimum 0.1mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. Can tolerate up to 15% TDS. MicroMist is the standard configuration for many ICP manufacturers due to its excellent sensitivity and precision at low flow rates. Replacement parts include P/Ns NFT-025 and 502703076.Recommend maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7363
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.