Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micropipette Tips, Low Retention, Bulk
SDPSCI_ED

Catalog No. S8538029 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
300 μL
1000 μL
Tip Style:
Beveled, Low Retention
Extended Length, Low Retention
Low Retention
UltraFine, Low Retention
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
S8538029 10 μL Low Retention
S8538030 10 μL Extended Length, Low Retention
S8538031 200 μL Beveled, Low Retention
S8538032 200 μL UltraFine, Low Retention
S8538033 300 μL Low Retention
S8538034 1000 μL Low Retention
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S8538029 Supplier United Scientific™ Supplier No. UNP10001
Add to Cart
Add to Cart

Low Retention Micropipette Tips are disposable tips intended for use with most brands of pipettors.

  • Deliver 10x lower retention levels with viscous samples, as compared to standard pipette tips.
  • Highly homogenous surface is free of defects, preventing non-specific binding.
  • Manufactured from ultra pure USP Class VI certified virgin polypropylene.
  • Free of DNase, RNase and pyrogen.
  • Natural color.
  • Bulk packaging.

Specifications

Description Low Retention Micropipette Tips
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Model UNP10001
Certifications/Compliance USP Class VI certified
For Use With (Application) Pipetting
Format Bagged
Tip Style Low Retention
Packaging Bagged
Quantity 1000 Pk.
Product Type Micropipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.