Learn More
Cytiva MICROPLATE FOIL, 384 WELL
Supplier:  Cytiva BR100577
Encompass
Microplate Foil 384-well for use in Biacore 4000, Biacore A100, Biacore T200, Biacore T100 and Biacore S51
Catalog No. 50-105-5335