Learn More
V&P Scientific Inc MICROPLATE SEALING SYSTEM, 3-Part, Fits all 96, 384, 1536 Standard Microplates
Supplier:  V&P Scientific Inc VP 416ALEMP
Encompass
MICROPLATE SEALING SYSTEM, 3-Part, for HTE, Parallel Synthesis and Reaction Optimization with SLAS Footprint Microplate Block, Lid and Tightening Knob, Hard Anodized Aluminum, Fits all 96, 384, 1536 Standard Microplates, Required PFA Films and Rubber Mat Sold Separately
Catalog No. NC2401892
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.