Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Microplates for Fluorescence-based Assays

Catalog No. M33090
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
96
384
Quantity:
10 Plates
30 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Quantity
M33090 96 30 Plates
M33089 96 10 Plates
M33091 384 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M33090 Supplier Invitrogen™ Supplier No. M33090
Add to Cart
Add to Cart

Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.

Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).

Features of the microplates for fluorescence-based assays include:

  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Content And Storage Store at room temperature.
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 30-plates
Product Line Invitrogen
Color Clear Bottom, Black
Disposable Single-use
Product Type Microplates
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 200 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) Nucleic Acid Quantification
Quantity 30 Plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.