Learn More
Description
- 3M™ Micropore™ Surgical Tape
- Part of the Multi Purpose Tape Category
- Offers gentleness to skin yet adheres appropriately for general securement needs.
Specifications
Specifications
| Product Type | Surgical Tape with Dispenser |
| Length (Metric) | 2.5 cm |
| Width (Metric) | 9.1 m |
| Quantity | 12 Rolls/Cs. |
| Dimensions (L x W) | 10 yd. x 1 in. |
| Model | 1535-1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.