Learn More
MicroGuard MP™ Microporous Coveralls with Attached Hood
Shop All International Enviroguard Systems ProductsDescription
- Comfortable state-of-the-art lamination technology allows for maximum breathability.
- Soft non-woven fabric combined with a microporous film allows moisture vapor to escape, so heat and moisture are not trapped near the body.
- Attached hood, along with elastic wrists and ankles, provide protection of the neck and shoulders and a secure fastening in high-movement areas where skin exposure can happen.
- Coveralls offer base-level protection against light liquid splashes and nonhazardous particulates providing the optimal balance of comfort, durability, and protection.
- Roomy, seamless shoulder area resists tears and provides more protection in key exposure areas.
- Wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric.
- Elastic back stretches with user for more comfort and better tear resistance.
- Elastic wrists and ankles offer a secure fit to help prevent skin exposure.
- Environmentally friendly, can be recycled after use.
- Attached hood provides additional protection of the neck and shoulders.
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 5X-Large |
| Gender | Unisex |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.