Learn More
International Enviroguard™ MicroGuard MP™ Microporous Lab Coats, No Pockets
Shop All International Enviroguard Systems ProductsDescription
- Microporous, disposable lab coats offer the balance of safety and comfort
- Fabric keeps dust and liquids out while allowing air to flow freely to keep worker cool
- Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
- Produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
- Breathable fabric lets moisture vapor escape
- No pockets
- Folded collar
- Environmentally friendly- can be recycled after use
- Elastic wrists offer a snug, secure fit to keep sleeves in place to prevent skin exposure
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Microporous Composite Fabric |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| No. of Pockets | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.