Learn More
International Enviroguard™ MicroGuard MP™ Microporous Lab Coats, No Pockets
Catalog No. 19472458 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-458 3X-Large
19-472-454 Medium
19-472-455 Large
19-472-456 X-Large
19-472-457 2X-Large
19-472-459 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-458 Supplier International Enviroguard™ Supplier No. 80463XL
Add to Cart
Add to Cart
Microporous lab coats deliver protection and comfort through a strong and durable fabric designed for worker comfort.
  • Microporous, disposable lab coats offer the balance of safety and comfort
  • Fabric keeps dust and liquids out while allowing air to flow freely to keep worker cool
  • Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed
  • Produced with state-of-the-art lamination technology to combine a soft non-woven fabric with a microporous film that repels liquids
  • Breathable fabric lets moisture vapor escape
  • No pockets
  • Folded collar
  • Environmentally friendly- can be recycled after use
  • Elastic wrists offer a snug, secure fit to keep sleeves in place to prevent skin exposure
Specifications
Product Type Controlled Environments Lab Coat
Size 3X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Folded
Seam Type Serged