Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valutek™ Microporous Lab Coats with Full Zipper

Catalog No. 191502094
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-2094 X-Large
19-150-2091 Small
19-150-2092 Medium
19-150-2093 Large
19-150-2095 -
19-150-2096 -
19-150-2097 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-150-2094 Supplier Valutek™ Supplier No. VTMLBCTZXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lab coats are manufactured with non-linting microporous film bonded with a non-woven fabric to provide barrier protection against non-toxic particulates, liquids, sprays, and dust.

  • Highly breathable and comfortable
  • Knee length with zipper closure, one pocket, elastic cuff, and a single-ply shirt collar design
  • Tested and manufactured in ISO compliant facilities to ensure quality standard and product specification compliance
  • Recommended for either stringent industrial or demanding cleanroom applications
  • Ideal for use in a cleanroom Class 1-10 (ISO 3-4) critical environment that requires high contamination control and cleanliness

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Collar Type Single Ply
For Use With (Application) Electronics, Medical Device, Pharmaceutical
Cleanroom Class Class 1-10 (ISO 3-4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.