Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Microprotein Reagents

Catalog No. p-318970 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
2 x 120mL Reagent, 1 x 15mL Standard
8 x 50mL Reagent, 1 x 15mL Standard
Formulation:
Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants and preservative, Protein standard solution: Contains Albumin 50mg/dL in Saline with Sodium Azide 0.05% as a preservative
Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants, preservative
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-376 2 x 120mL Reagent, 1 x 15mL Standard Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants and preservative, Protein standard solution: Contains Albumin 50mg/dL in Saline with Sodium Azide 0.05% as a preservative
23-666-375 8 x 50mL Reagent, 1 x 15mL Standard Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants, preservative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-376 Supplier Pointe Scientific Supplier No. P7582240
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Microprotein Reagent includes: Buffer; Pyrogallol Red 0.067mM; Sodium molybdate stabilizer 0.153mM; surfactants; preservative
  • Protein Standard Solution includes: Ablumin 50mg/dL in saline; Sodium azide 0.05%

Quantify total protein levels in urine

  • Method: Pyragallol Red, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 600nm
  • Linearity: 250mg/dL
  • Expected Values: 28 to 141mg (24 hr.)

For the quantitative determination of total protein in urine

Specifications

Certifications/Compliance CE Marked
Type Microprotein Reagent Kit
Formulation Microprotein reagent: Contains Buffer, Pyrogallol Red 0.067mM/L, Sodium Molybdate Stabilizer 0.153mM/L, Surfactants and preservative, Protein standard solution: Contains Albumin 50mg/dL in Saline with Sodium Azide 0.05% as a preservative
For Use With (Application) Quantitative determination of total protein in urine
Quantity 2 x 120mL Reagent, 1 x 15mL Standard
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.