Learn More
ATCC MicroQuant™ Salmonella enterica, low CFU (ATCC 14028-LQ-PACK™)
Supplier:  ATCC 14028LQPACK
Encompass
MicroQuant™ Salmonella enterica (low CFU) is a quantitated, best-in-class microbial control. This ISO 17034 reference material is provided as a pack containing 5 vials of cryopreserved pellets (100 to 1,000 CFU per vial) and 5 vials of rehydration buffer. This single-use format is stored at 2–8°C and rehydrates rapidly. The strain is recommended for use in compendial testing.
Biosafety Level: BSL-2
Find more MicroQuant™ products
Catalog No. NC3853591