Learn More
Cliawaived Inc Microsafe 75 uL Tubes (50 per Bag)
SDP
Supplier:  Cliawaived Inc ST107550
Encompass_Preferred
(50 Tubes, 75 uL)Microsafe Tube One piece collection and dispensing tubes. Ideal for fingerstick sampling and dispensing. For In vitro use. Available in sizes 15uL, 20 uL, 40uL, and 75uL.
Catalog No. NC1859640
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.