Learn More
Perkin Elmer US LLC Microsample Cup for the Spectrum One/100/400/65
Supplier:  Perkin Elmer US LLC L1201655
Encompass_Preferred
PerkinElmer's Diffuse Reflectance Accessory comes with abrasive sampling tools that allow you to collect ready-to-analyze dust samples from hard-surfaced solid materials. The Diffuse Reflectance Multi-Purpose Sample Holder (L1201865), designed to hold abrasive sticks, pads, or sample cups, easily slides into the diffuse reflectance accessory.
Catalog No. 50-211-3391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.