Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Microsart™ AMP Extraction Kit

Catalog No. 14577841 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
50 Columns/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-577-841 50 Columns/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-577-841 Supplier Sartorius Supplier No. SMB952003
Add to Cart
Add to Cart

For use with Microsart™ Mycoplasma AMP, Research quantitative detection kits.

  • Used for isolation of genomic DNA from different types of sample material
  • Compatible with Microsart™ Mycoplasma AMP, Research qPCR kits
  • Sensitive and robust detection of Mycoplasma within test samples

Compliance

EP 2.6.7

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Sample Type Cell Culture Supernatants
For Use With (Application) Mycoplasma Detection
Includes Buffers, collection tubes, sample storage tubes
Product Type Extraction Kit
Quantity 50 Columns/Kit
Sample Volume 200μL
Species Mycoplasma
Target DNA
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.