Learn More
Description
- Ready-to-use agar media dish (prefilled with different agar media types) with active lid
- Sterile packaged and ready to use together with Microsart filter units
- Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
- Liftable interior lid provides easy access to select colonies after incubation for further analysis
Specifications
Specifications
| Classification | Animal Origin |
| Form | Gel |
| Product Type | Growth Medium |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Sterilization Method | Gamma-irradiated |
| With Additives | BPE |
| Without Additives | No Cholesterol |
| For Use With (Application) | Target microorganisms: total count |
| Formulation | Solid Agar Medium |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.