Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MicroScan™ QC Sets

Catalog No. 22246501
Change view
Click to view available options
Quantity:
5 Loops/Pack
5 Loops/Pk.
6 Loops/Set
8 Loops/Set
Product Type:
Dried Gram Neg QC 1 Loop Set
Gram Neg QC Set
Gram Pos QC 1 Loop Set
Gram Pos QC Set
HNID QC Set
Rapid Anaerobe ID QC Set
Rapid Gram Neg QC Set
Rapid Yeast ID QC Set
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-246-501 8 Loops/Set Gram Pos QC 1 Loop Set
R4661001 5 Loops/Pack Gram Pos QC Set
R4661000 5 Loops/Pack Gram Neg QC Set
R4661002 5 Loops/Pk. Rapid Gram Neg QC Set
R4661003 5 Loops/Pack Rapid Anaerobe ID QC Set
R4661004 5 Loops/Pack HNID QC Set
R4661005 5 Loops/Pk. Rapid Yeast ID QC Set
22-246-500 6 Loops/Set Dried Gram Neg QC 1 Loop Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-246-501 Supplier Thermo Scientific™ Supplier No. R4681001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.

Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ MicroScan™ QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.

Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.

Convenience

  • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.

Performance

  • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.

Value

  • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.

Confidence

  • Reach complete traceability with ATCC™ License Derivative strains.

Efficiency

  • Achieve accurate and convenient documentation with peel-off, transferable labels.

Safety

  • Reduce the risk of contamination and infection with zero organism handling.
  • More than 600 quality control organisms, including convenient sets for commonly used identification systems
  • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
  • ATCC™ Licensed Derivative Program member

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC™. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 29212TM*, 51299™*, 35218™*, 49732™*, 29213™*, 43300™*, BAA-977™*, 49147™*
Certifications/Compliance ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
Content And Storage 2°C to 8°C
Format Culti-Loops
Product Type Gram Pos QC 1 Loop Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Packaging Type 8 Loops/Set
Quantity 8 Loops/Set
Species Enterococcus faecalis, Escherichia coli, Micrococcus luteus, Staphylococcus aureus subsp. aureus, Streptococcus gallolyticus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.