Learn More
Description
- Eyepiece for microscope
Warranty and Services
1 Year
Specifications
Specifications
| Type | Eyepiece |
| Field of View | 11 mm |
| Magnification Power | 20X |
| Size | Tube size: 23.2 mm |
| Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
