Learn More
Eisco™ Microscope Glass Slides
SCI_ED
Catalog No. S24460 Shop All Eisco Products
Change view
Click to view available options
Quantity:
72/Pk.
Catalog No. Quantity
S24460 72/Pk.
1 options
Catalog No. S24460 Supplier Eisco™ Supplier No. BI0084B
Add to Cart
Add to Cart
Made from clear and optically-flat glass.
  • Frosted glass
  • Ground and polished edges
Specifications
Dimensions (L x W) 75 x 25 mm
For Use With (Equipment) Ground and polished edges
Material Clear, Optically-flat Glass
Quantity 72/Pk.
Width (Metric) 1.1 to 1.2 mm thick
Product Type Microscope Glass Slides