Learn More
United Scientific™ Microscope Slide Box, ABS
SDPSCI_ED
Catalog No. S012568 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Blue
White
Slide Capacity:
25
50
100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Slide Capacity
S012568 Blue 100
S012573 White 25
S012572 Blue 25
S012571 White 50
S012570 Blue 50
S012569 White 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S012568 Supplier United Scientific™ Supplier No. 62612
Add to Cart
Add to Cart
Classic design to safely store microscope slides.
  • Designed as a convenient solution for temporary storage and transferring of microscope slides.The bottom is lined with a cork pad, and the lid has a printed index for quick reference.
  • Accommodates microscope slides in two sizes:
    • 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
    • 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
  • Made of strong ABS material
  • Snap closures and hinged-lids provide safe operation
  • A water-proof index card is mounted inside the lids for simple specimen identification
  • Cork sheet protects the slides
  • The 100-place slide box includes a metal snap closure
Specifications
Type Slide Storage Box
Slide Capacity 100
Color Blue
Material ABS