Description
- Designed as a convenient solution for temporary storage and transferring of microscope slides.The bottom is lined with a cork pad, and the lid has a printed index for quick reference.
- Accommodates microscope slides in two sizes:
- 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
- 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
- Made of strong ABS material
- Snap closures and hinged-lids provide safe operation
- A water-proof index card is mounted inside the lids for simple specimen identification
- Cork sheet protects the slides
- The 100-place slide box includes a metal snap closure
Specifications
Specifications
| Type | Slide Storage Box |
| Slide Capacity | 50 |
| Color | Blue |
| Material | ABS |