Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lieder™ Microscope Slide School Sets A, B, C and D
SCI_ED

Catalog No. S041399 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Quantity:
175 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S041399 175 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S041399 Supplier Lieder™ Supplier No. 850
Add to Cart
Add to Cart
This item is not returnable. View return policy

Basic component of the Multimedia Program For Microscopic Biology comprising of 175 microscope slides. These sets contain slides of typical microorganisms, cell division and embryonic development, as well as tissues or organs of plants, animals and man.

The Mutimedia Program For Microscopic Biology aims to give a strictly outlined synopsis of all lines of biology important for instruction at schools, colleges and universities suitable for working with the microscope. A considerable part of the Program is an extensive manual with detailed descriptions and drawings of the prepared microscope slides and photomicrographs of the school series A, B, C and D. Well-selected complementary media package with overhead transparencies, sketch and work sheets, descriptions and pictures of drawings, and new CD-ROM serves teacher to work with the subject during the lessons. Abundant material offers teacher opportunity to select and vary content to tailor lessons for each class.

  • Four series sets are arranged systematically and constructively compiled so that each enlarges the subject line of the proceeding one
  • Each of the slides has been carefully selected on the basis of its instructional value

Specifications

Format Class Set
Quantity 175 Slides
Languages English, French, German, Spanish
Content And Storage Ambient Temperature
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.