Learn More
Simport™ Scientific Microscreen™ I Small Biopsy Processing/Embedding Cassettes
Shop All Simport Scientific ProductsDescription
- Microscreen™ cassette is made of a strong nylon mesh
- Designed to handle small, fragile biopsies
- Nylon mesh has 1,500 square openings (300 μm each)
- 8 x 11 mm slots in base and lid to help cassette sink rapidly
- Sturdy nylon mesh improves fluid exchange by reducing surface tension of reagents
- Accelerates fixation/preparation of tissue samples
- Eliminates the need for pads, sponges, nylon or lens paper bags
- Reduces the number of incidents involving non-fixed or insufficiently fixed tissues
- Inner chamber design for greater biopsy protection
- Large front tab for easier lid opening
- Hinge design configuration allows for multiple opening and closing
Specifications
Specifications
| Writing Face Angle | 45° |
| Color | Orange |
| Material | Acetal |
| Product Type | Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.