Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Microscreen™ I Small Biopsy Processing/Embedding Cassettes

Catalog No. 23045395 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Orange
Peach
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-395 Peach
23-045-388 Orange
23-045-393 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-045-395 Supplier Simport™ Scientific Supplier No. M5207
This item is not returnable. View return policy

Microscreen™ I Small Biopsy Processing/Embedding Cassettes are designed for small and delicate specimens.

  • Microscreen™ cassette is made of a strong nylon mesh
  • Designed to handle small, fragile biopsies
  • Nylon mesh has 1,500 square openings (300 μm each)
  • 8 x 11 mm slots in base and lid to help cassette sink rapidly
  • Sturdy nylon mesh improves fluid exchange by reducing surface tension of reagents
  • Accelerates fixation/preparation of tissue samples
  • Eliminates the need for pads, sponges, nylon or lens paper bags
  • Reduces the number of incidents involving non-fixed or insufficiently fixed tissues
  • Inner chamber design for greater biopsy protection
  • Large front tab for easier lid opening
  • Hinge design configuration allows for multiple opening and closing

Specifications

Writing Face Angle 45°
Color Peach
Material Acetal
Product Type Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.