Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroSEQ™ 500 16S rDNA PCR Kit
GREENER_CHOICE

Catalog No. 4348228
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-482-28 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-482-28 Supplier Applied Biosystems™ Supplier No. 4348228
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MicroSEQ™ 500 16S rDNA PCR Kit is the PCR component of the MicroSEQ™ 500 16S rDNA Bacterial Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria. PCR-amplified products generated with this kit can be sequenced using the MicroSEQ™ 500 16S rDNA Sequencing Kit.

The MicroSEQ™ 500 16S rDNA PCR Kit is the PCR component of the MicroSEQ™ 500 16S rDNA Bacterial Identification System, which provides an easy-to-use DNA sequence-based method to identify most bacteria. PCR-amplified products generated with this kit can be sequenced using the MicroSEQ™ 500 16S rDNA Sequencing Kit. The two kits together permit the quick and convenient amplification and sequencing of 500 base pairs of the 16S rRNA gene, which can then be compared to the MicroSEQ™ 16S rDNA 500 Library to obtain an identity match. This kit does not include a copy of the protocol, nor a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-same work flow as the MicroSEQ D2 LSU fungal kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution

Streamlining every step in microbial identification, Applied Biosystems MicroSeq System combines the advantages of MicroSEQ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSeq bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Specifications

Content And Storage Each kit contains enough PCR Master Mix to perform 50 PCR amplifications, 5 negative control assays, and 5 positive control assays. One tube of positive control E. coli DNA, and one of water (for use as a negative control) are also included. Store at -15 to -25°C upon reciept. After first use, store at 2–8 °C in a PCR Clean Room.
Detection Method Primer-probe
Technique PCR Amplification, SBT (Sequence-Based Typing)
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler
Libraries Customizable, Bacterial Library (> 2000 species)
Product Line MicroSEQ
Product Type PCR Kit
Quantity 1 Kit
Shipping Condition Dry Ice
Software MicroSEQ™ ID Analysis Software
Target 1st 500 bp of 16S rRNA gene
Target Organism Class Bacteria
For Use With (Application) Microbial Detection
Sample Type DNA (Genomic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.