Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroSEQ™ Full Gene 16S rDNA PCR Kit
GREENER_CHOICE

Catalog No. 4349155
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-491-55 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-491-55 Supplier Applied Biosystems™ Supplier No. 4349155
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MicroSEQ Full Gene 16S rDNA PCR Kit is the PCR component of the MicroSEQ 16S rDNA Full Gene Identification Kit, which provides an easy-to-use DNA sequence-based method to identify most bacteria.

The MicroSEQ™ Full Gene 16S rDNA PCR Kit is the PCR component of the MicroSEQ™ 16S rDNA Full Gene Identification Kit, which provides an easy-to-use DNA sequence-based method to identify most bacteria. Use the PCR kit to amplify the bacterial 16S rRNA gene, for subsequent sequencing using the MicroSEQ™ Full Gene 16S rDNA Sequencing Kit. This PCR kit does not include a protocol or a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-similar work flow as the MicroSEQ™ D2 LSU fungal kit and the MicroSeq 500 16S rDNA kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution

Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Specifications

Content And Storage • 2X PCR Master Mix (sufficient for 20 amplifications), 3 tubes
• Positive Control DNA, 1 tube
• Negative Control DNA (water), 1 tube
• Protocol and Quick Reference Guide

After first use, store at 2°C to 8°C in a PCR clean room.
Detection Method Primer-probe
GC-Rich PCR Performance Low
Technique SBT (Sequence-Based Typing)
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler
Libraries Customizable, Bacterial Library (> 2000 species)
Product Line MicroSEQ
Product Type Full Gene 16S rDNA PCR Kit
Quantity 1 Kit
Shipping Condition Dry Ice
Software MicroSEQ™ ID Analysis Software
Target 16S rRNA (full gene)
Target Organism Class Bacteria
For Use With (Application) Standard PCR
Sample Type DNA (Genomic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.