Learn More
Applied Biosystems™ MicroSEQ™ ID 16S rDNA 500 Supplemental Library, v2019
Description
The MicroSEQ ID 16S rDNA 500 Supplemental Library v2019 is a stand-alone library that extends the size of the MicroSEQ ID 16S rDNA 500 Library v2019 to strengthen the identification process when using MicroSEQ ID Microbial Identification Software. The Supplemental library contains 7,138 16s rDNA public sequences of type and non-type strains of bacterial species that underwent the same rigorous validation process as the sequences in the MicroSEQ ID 16S rDNA 500 Library v2019. The Supplemental Library is used when the MicroSEQ ID 16S rDNA 500 Library v2019 results in a percent match lower than the species ID acceptance criteria.
Key features include:
- In-silico validated library with over 7,000 type and non-type strains quality-checked for sequence, taxonomy, nomenclature, and phylogeny
- Reliable entries sourced from public resources converted into a simple format for routine validation of unique environmental isolates
Specifications
Specifications
| Quantity | Each |
| Product Line | MicroSEQ |
| For Use With (Equipment) | MicroSEQ™ ID 16S rDNA 500 Library v2019 |
| Product Type | Supplemental Library |
| Shipping Condition | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.